missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine BTC Protein
A cDNA sequence encoding the BTC was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 61725.00 SEK
Specifications
Gene ID (Entrez) | 280737 |
---|---|
Name | BTC Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | BTC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15964188
|
enQuireBio™
QP10540-5UG |
5 μg |
1545.00 SEK
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954188
|
enQuireBio™
QP10540-20UG |
20 μg |
2405.00 SEK
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944188
|
enQuireBio™
QP10540-1MG |
1 mg |
61725.00 SEK
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
280737 | |
Research Use Only | |
BTC | |
Bovine | |
Untagged | |
The Betacellulin Bovine Recombinant was lyophilized after extensive dialysis against 50mM acetic acid. |
BTC Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MARAAPGSGA SPLPLLPALA LGLVILHCVV ADGNSTRSPE DDGLLCGDHA ENCPATTTQP KRRGHFSRCP KQYKHYCIKG RCRFVVAEQT PSCVCDEGYA GARCERVDLF YLRGDRGQIL VICLIAVMVI FIILVVSICT CCHPLRKRRK RRKKEEEMET LGKDITPIND DIQETSIA | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |