missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Cathelicidin-4 Protein
A DNA sequence encoding the Bos taurus (Bovine) Cathelicidin-4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7211-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_777252.1 | |
Cathelicidin-4 Protein | |
Research Use Only | |
CATHL4 | |
Recombinant Protein | |
E. coli | |
ILPWKWPWWP WRR | |
Greater than 90% as determined by SDS-PAGE. |
282166 | |
500 μg | |
Not determined. | |
Full Length | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |