missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Cathelicidin-4 Protein

A DNA sequence encoding the Bos taurus (Bovine) Cathelicidin-4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7211-ec-50ug

 View more versions of this product

Product Code. 16047730

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_777252.1
Cathelicidin-4 Protein
Research Use Only
CATHL4
Recombinant Protein
E. coli
ILPWKWPWWP WRR
Greater than 90% as determined by SDS-PAGE.
282166
50 μg
Not determined.
Full Length
Bovine
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers