missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Hemagglutinin-esterase Protein

A DNA sequence encoding the Bos taurus (Bovine) Hemagglutinin-esterase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9549-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16058461

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Hemagglutinin-esterase Protein
Research Use Only
HE
Recombinant Protein
Yeast
FDNPPTNVVS HLNGDWFLFG DSRSDCNHVV NTNPRNYSYM DLNPALCDSG KISSKAGNSI FRSFHFTDFY NYTGEGQQII FYEGVNFTPY HAFKCTTSGS NDIWMQNKGL FYTQVYKNMA VYRSLTFVNV PYVYNGSAQS TALCKSGSLV LNNPAYIARE ANFGDYYYKV EADFYLSGCD EYIVPLCIFN GKFLSNTKYY DDSQYYFNKD TGVIYGLNST ETITTGFDFN CHYLVLPSGN YLAISNELLL TVPTKAICLN KRKDFTPVQV VDSRWNNARQ SDNMTAVACQ PPYCYFRNST TNYVGVYDIN HGDAGFTSIL SGLLYDSPCF SQQGVFRYDN VSSVWPLYSY GRCPTAADIN TPDVPICVYD PLPLILLGIL LGVAVIIIVV LLLYFMVDNG TRLHDA
Greater than 81.30% as determined by SDS-PAGE.
1 mg
Not determined.
Full Length
Bovine
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers