missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Inhibin alpha chain Protein
A DNA sequence encoding the Bos taurus (Bovine) Inhibin alpha chain, was expressed in the hosts and tags indicated.
3180.00 SEK - 38570.00 SEK
Specifications
Accession Number | NP_776519.2 |
---|---|
Gene ID (Entrez) | 281254 |
Name | Inhibin alpha chain Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16051780
|
enQuireBio™
QP6231-EC-100UG |
100 μg |
11125.00 SEK
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071780
|
enQuireBio™
QP6231-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16061780
|
enQuireBio™
QP6231-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081780
|
enQuireBio™
QP6231-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091780
|
enQuireBio™
QP6231-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001790
|
enQuireBio™
QP6231-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_776519.2 | |
Inhibin alpha chain Protein | |
Not determined. | |
Full Length | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
281254 | |
Research Use Only | |
INHA | |
Recombinant Protein | |
E. coli | |
STPPLPWPWS PAALRLLQRP PEEPAAHADC HRAALNISFQ ELGWDRWIVH PPSFIFYYCH GGCGLSPPQD LPLPVPGVPP TPVQPLSLVP GAQPCCAALP GTMRPLHVRT TSDGGYSFKY EMVPNLLTQH CACI | |
Greater than 90% as determined by SDS-PAGE. |