missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Metalloproteinase inhibitor 4 Protein
A DNA sequence encoding the Bos taurus (Bovine) Metalloproteinase inhibitor 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6796-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001039336.1 | |
Metalloproteinase inhibitor 4 Protein | |
Research Use Only | |
TIMP4 | |
Recombinant Protein | |
E. coli | |
CSCAPAHPQQ HVCHSALAIR AKISSEKVVP ASTDPADPQK MIRYEIKQIK MFKGFEKVND IQYIYTPFDS SLCGVKLEAN SQKRYLLTGQ ILSDGKVFVH LCNYIEPWEN LSFLQRESLN HHYHLNCGCQ ITTCYAVPCT ISAPNECLWT DWLLERKLYG YQAQHYVCMK HVDGSCSWYQ GRLPLRKEFV DIIQP | |
Greater than 90% as determined by SDS-PAGE. |
317694 | |
200 μg | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |