missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Pregnancy-associated glycoprotein 1
A DNA sequence encoding the Bos taurus (Bovine) Pregnancy-associated glycoprotein 1, was expressed in the hosts and tags indicated.
7405.00 SEK - 20560.00 SEK
Specifications
Accession Number | NP_776836.1 |
---|---|
Gene ID (Entrez) | 281964 |
Name | Pregnancy-associated glycoprotein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16069950
|
enQuireBio™
QP7639-BA-20UG |
20 μg |
7405.00 SEK
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16059950
|
enQuireBio™
QP7639-BA-100UG |
100 μg |
20560.00 SEK
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16079950
|
enQuireBio™
QP7639-BA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_776836.1 | |
Pregnancy-associated glycoprotein 1 | |
Not determined. | |
Full Length of Mature Protein | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |
281964 | |
Research Use Only | |
PAG1 | |
Recombinant Protein | |
Baculovirus | |
RGSNLTTHPL RNIKDLVYMG NITIGTPPQE FQVVFDTASS DLWVPSDFCT SPACSTHVRF RHLQSSTFRL TNKTFRITYG SGRMKGVVVH DTVRIGNLVS TDQPFGLSIE EYGFEGRIYD GVLGLNYPNI SFSGAIPIFD KLKNQRAISE PVFAFYLSKD EREGSVVMFG GVDHRYYEGE LNWVPLIQAG DWSVHMDRIS IERKIIACSD GCKALVDTGT SDIVGPRRLV NNIHRLIGAI PRGSEHYVPC SEVNTLPSIV FTINGINYPV PGRAYILKDD RGRCYTTFQE NRVSSSTETW YLGDVFLRLY FSVFDRGNDR IGLARAV | |
Greater than 88.1% as determined by SDS-PAGE. |