missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine TGF-beta 1 / TGFB1 Protein

A DNA sequence encoding the Bos taurus (Bovine) TGF-beta 1 / TGFB1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6778-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16065780

  • 12765.00 SEK / 200µg


Estimated Shipment
02-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_001159540.1
TGF-beta 1 / TGFB1 Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Bovine
His
Tris-based buffer, 50% glycerol
282089
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TGFB1
Recombinant Protein
E. coli
ALDTNYCFSS TEKNCCVRQL YIDFRKDLGW KWIHEPKGYH ANFCLGPCPY IWSLDTQYSK VLALYNQHNP GASAAPCCVP QALEPLPIVY YVGRKPKVEQ LSNMIVRSCK CS
Videos
SDS
Documents