missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine TNF-alpha Protein
A DNA sequence encoding the Bos taurus (Bovine) TNF-alpha, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8605-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_776391.2 | |
TNF-alpha Protein | |
Research Use Only | |
TNF | |
Recombinant Protein | |
E. coli | |
LRSSSQASSN KPVAHVVADI NSPGQLRWWD SYANALMANG VKLEDNQLVV PADGLYLIYS QVLFRGQGCP STPLFLTHTI SRIAVSYQTK VNILSAIKSP CHRETPEWAE AKPWYEPIYQ GGVFQLEKGD RLSAEINLPD YLDYAESGQV YFGIIAL | |
Greater than 90% as determined by SDS-PAGE. |
280943 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Bovine | |
GST | |
Tris-based buffer, 50% glycerol |