missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine TNF-alpha Protein
A DNA sequence encoding the Bos taurus (Bovine) TNF-alpha, was expressed in the hosts and tags indicated.
3070.00 SEK - 27205.00 SEK
Specifications
Accession Number | NP_776391.2 |
---|---|
Gene ID (Entrez) | 280943 |
Name | TNF-alpha Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16025531
|
enQuireBio™
QP8605-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065531
|
enQuireBio™
QP8605-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16015531
|
enQuireBio™
QP8605-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045531
|
enQuireBio™
QP8605-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16055531
|
enQuireBio™
QP8605-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16035531
|
enQuireBio™
QP8605-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_776391.2 | |
TNF-alpha Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Bovine | |
GST | |
Tris-based buffer, 50% glycerol |
280943 | |
Research Use Only | |
TNF | |
Recombinant Protein | |
E. coli | |
LRSSSQASSN KPVAHVVADI NSPGQLRWWD SYANALMANG VKLEDNQLVV PADGLYLIYS QVLFRGQGCP STPLFLTHTI SRIAVSYQTK VNILSAIKSP CHRETPEWAE AKPWYEPIYQ GGVFQLEKGD RLSAEINLPD YLDYAESGQV YFGIIAL | |
Greater than 90% as determined by SDS-PAGE. |