missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. avellana 2S albumin Protein
A DNA sequence encoding the Corylus avellana 2S albumin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6921-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
2S albumin Protein | |
Research Use Only | |
Full Length | |
C. avellana | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
200 μg | |
Not determined. | |
Recombinant Protein | |
E. coli | |
FRTTITTVDV DEDIVNQQGR RGESCREQAQ RQQNLNQCQR YMRQQSQYGS YDGSNQQQQQ ELEQCCQQLR QMDERCRCEG LRQAVMQQQG EMRGEEMREV METARDLPNQ CRLSPQRCEI RSARF | |
Greater than 90% as determined by SDS-PAGE. |