missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. botulinum Botulinum neurotoxin type B Protein
A DNA sequence encoding the Clostridium botulinum Botulinum neurotoxin type B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9517-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
Botulinum neurotoxin type B Protein | |
Research Use Only | |
botB | |
Recombinant Protein | |
Yeast | |
MPVTINNFNY NDPIDNNNII MMEPPFARGT GRYYKAFKIT DRIWIIPERY TFGYKPEDFN KSSGIFNRDV CEYYDPDYLN TNDKKNIFLQ TMIKLFNRIK SKPLGEKLLE MIINGIPYLG DRRVPLEEFN TNIASVTVNK LISNPGEVER KKGIFANLII FGPGPVLNEN ETIDIGIQNH FASREGFGGI MQMKFCPEYV SVFNNVQENK GASIFNRRGY FSDPALILMH ELIHVLHGLY GIKVDDLPIV PNEKKFFMQS TDAIQAEELY TFGGQDPSII TPSTDKSIYD KVLQNFRGIV DRLNKVLVCI SDPNININIY KNKFKDKYKF VEDSEGKYSI DVESFDKLYK SLMFGFTETN IAENYKIKTR ASYFSDSLPP VKIKNLLDNE IYTIEEGFNI SDKDMEKEYR GQNKAINKQA YEEISKE | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
C. botulinum | |
His | |
Tris-based buffer, 50% glycerol |