missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. botulinum clpP Protein

A DNA sequence encoding the Clostridium botulinum (strain Hall / ATCC 3502 / NCTC 13319 / Type A) clpP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7429-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16018830

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

WP_003357557.1
clpP Protein
10 μg
Not determined.
Full Length
C. botulinum
His-SUMO
Tris-based buffer, 50% glycerol
5187372
Greater than 90% as determined by SDS-PAGE.
Research Use Only
clpP
Recombinant Protein
E. coli
MSLVPVVVEQ TNRGERSYDI YSRLLKDRII MLSEEVNDTT ASLIVAQLLF LEAEDPDKDI HLYINSPGGS ITSGMAIYDT MQYIKPDVST ICVGMAASMG AFLLAAGAKG KRYALPNSEV MIHQPLGGFR GQATDIGIHA ERILKMKKKL NTILSDRTGK PLEQVELDTE RDHFLSAEEA KEYGLIDEVI DKKK
Videos
SDS
Documents
Certificates
Special Offers

Special Offers