Learn More
enQuireBio™ Recombinant C. botulinum Flagellin Protein
A DNA sequence encoding the Clostridium botulinum (strain Hall / ATCC 3502 / NCTC 13319 / Type A) Flagellin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6926-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
WP_011987021.1 | |
Flagellin Protein | |
1 mg | |
Not determined. | |
Full Length | |
C. botulinum | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5204320 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
CBO2730 | |
Recombinant Protein | |
E. coli | |
MIINHNLNAM NAHRQMAINT GNNGKAIEKL SSGLRINRAG DDAAGLAISE KMRGQIRGLN QASRNSQDAI SLIQTAEGAL NETHSILQRM RELTVQAAND TNVTVDRENL QKEVAELQSE INRISSQTQF NTKSLINGAV SKDADAMTFQ IGANSGQTIK LTIGSMDAKT LNVDKIKLDD TAANVTKQIA TIDNAINSVS KERAKLGANQ NRLEHTIANL DNSAENLQAA ESRIRDVDMA KEMMNFTKTN ILTQAAQAML AQANQAPQGV LQLLR |