missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. trachomatis Major outer membrane porin Protein

A DNA sequence encoding the Chlamydia trachomatis serovar L2 (strain 434/Bu / ATCC VR-902B) Major outer membrane porin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7292-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16058030

  • 7610.00 SEK / 100µg


Estimated Shipment
09-02-2021
Add to basket

Description

Description

Specifications

Specifications

WP_009873292.1
Major outer membrane porin Protein
100 μg
Not determined.
Full Length
C. trachomatis
His-SUMO
Tris-based buffer, 50% glycerol
5858320
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ompA
Recombinant Protein
E. coli
LPVGNPAEPS LMIDGILWEG FGGDPCDPCT TWCDAISMRM GYYGDFVFDR VLQTDVNKEF QMGAKPTTAT GNAAAPSTCT ARENPAYGRH MQDAEMFTNA AYMALNIWDR FDVFCTLGAT SGYLKGNSAS FNLVGLFGDN ENHATVSDSK LVPNMSLDQS VVELYTDTTF AWSAGARAAL WECGCATLGA SFQYAQSKPK VEELNVLCNA AEFTINKPKG YVGQEFPLDL KAGTDGVTGT KDASIDYHEW QASLALSYRL NMFTPYIGVK WSRASFDADT IRIAQPKSAT TVFDVTTLNP TIAGAGDVKA SAEGQLGDTM QIVSLQLNKM KSRKSCGIAV GTTIVDADKY AVTVETRLID ERAAHVNAQF RF
Videos
SDS
Documents