missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Cat Allergen Fel d 4 Protein
A DNA sequence encoding the Felis catus (Cat) (Felis silvestris catus) Allergen Fel d 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7729-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001009233.1 | |
Allergen Fel d 4 Protein | |
Research Use Only | |
LOC493739 | |
Recombinant Protein | |
E. coli | |
HEEENVVRSN IDISKISGEW YSILLASDVK EKIEENGSMR VFVEHIKALD NSSLSFVFHT KENGKCTEIF LVADKTKDGV YTVVYDGYNV FSIVETVYDE YILLHLLNFD KTRPFQLVEF YAREPDVSQK LKEKFVKYCQ EHGIVNILDL TEVDRCLQAR GSEVAQDSSV E | |
Greater than 90% as determined by SDS-PAGE. |
493739 | |
500 μg | |
Not determined. | |
Full Length | |
Cat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |