missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken MDH1 Protein
A cDNA sequence encoding the MDH1 was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 16985.00 SEK
Specifications
Name | MDH1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Chicken |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15941159
|
enQuireBio™
QP12665-1000IU |
1000 IU |
1545.00 SEK
1000 units |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951159
|
enQuireBio™
QP12665-50000IU |
50000 IU |
16985.00 SEK
50000 units |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15961159
|
enQuireBio™
QP12665-5000IU |
5000 IU |
2405.00 SEK
5000 units |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Spezifikation
MDH1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Chicken | |
Untagged | |
Each mg of protein contains 0.59 mg NaPO4. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGEPIRVLVT GAAGQIAYSL LYSIAKGDVF GKEQPLVLVL LDITPMMTVL EGVVMELQDC ALPLLREVIP TDKEEVAFKD LDIAILVGSM PRREGMERKD LLKANVKIFK SQGAALDKYA KKTVKVVVVG NPANTNCLIA SKSAPSIPKE NFSCLTRLDH NRAKSQIALK LGVTSNDVKN VIIWGNHSST QYPDVNHAKV NVKGKEVGVY EAIKDDSWLK GDFILTVQQR GAAVIKARKL SSAMSAAKAI CDHVRDIWFG TPAGEFVSMG VISDGNSYGV PEDLLYSFPV VIKDKTWKFV EGLPINDFSR EKMDLTAKEL TEEKETAVEF LSSA | |
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |