missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chicken Mitochondrial Rho GTPase 1 Protein

A DNA sequence encoding the Gallus gallus (Chicken) Mitochondrial Rho GTPase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7700-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16060281

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001006208.1
Mitochondrial Rho GTPase 1 Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Chicken
His-SUMO
Tris-based buffer, 50% glycerol
417410
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RHOT1
Recombinant Protein
E. coli
MKKDVRILLV GEPRVGKTSL IMSLVSEEFP EEVPPRAEEI TIPADVTPER VPTHIVDYSE AEQNDEQLYH EISQANVICI VYAVNNKNSI DKVTSRWIPL INERTDKDSR LPLILVGNKS DLVEYSSMET ILPIMNQYTE IETCVECSAK NLKNRSELFY YAQKAVLHPT GPLYCPEEKE MKPACIKALT RIFRISDQDN DGTLNDAELN FFQRICFNT
Videos
SDS
Documents