missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chinese Hamster Peroxiredoxin 1 / PRDX1 Protein

A DNA sequence encoding the Cricetulus griseus (Chinese hamster) (Cricetulus barabensis griseus) Peroxiredoxin 1 / PRDX1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8117-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16062761

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001233694.1
Peroxiredoxin 1 / PRDX1 Protein
500 μg
Not determined.
Full Length
Chinese Hamster
His
Tris-based buffer, 50% glycerol
100689332
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PRDX1
Recombinant Protein
E. coli
MSSGNAKIGY PAPNFKATAV MPDGQFRDIC LSEYRGKYVV FFFYPLDFTF VCPTEIIAFS DRAEEFKKLN CQVIGASVDS HFCHLAWINT PKKQGGLGPM NIPLVSDPKR TIAQDYGVLK ADEGISFRGL FIIDDKGILR QITINDLPVG RSVDEILRLV QAFQFTDKHG EVCPAGWKPG SDTIKPDVQK SKEYFSKQK
Videos
SDS
Documents