Learn More
enQuireBio™ Recombinant Cytomegalovirus 65 kDa phosphoProtein
A DNA sequence encoding the Human cytomegalovirus (strain AD169) (HHV-5) (Human herpesvirus 5) 65 kDa phosphoprotein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7365-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
65 kDa phosphoProtein | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Cytomegalovirus | |
His | |
Tris-based buffer, 50% glycerol |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
UL83 | |
Recombinant Protein | |
E. coli | |
FDIDLLLQRG PQYSEHPTFT SQYRIQGKLE YRHTWDRHDE GAAQGDDDVW TSGSDSDEEL VTTERKTPRV TGGGAMAGAS TSAGRKRKSA SSATACTSGV MTRGRLKAES TVAPEEDTDE DSDNEIHNPA |