missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog Cationic trypsin Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Cationic trypsin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9998-iv-50ug
Additional Details : Weight : 0.01000kg
Specifications
Cationic trypsin Protein | |
Research Use Only | |
Full Length | |
Dog | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
50 μg | |
Not determined. | |
Recombinant Protein | |
E. coli | |
IVGGYTCSRN SVPYQVSLNS GYHFCGGSLI NSQWVVSAAH CYKSRIQVRL GEYNIAVSEG GEQFINAAKI IRHPRYNANT IDNDIMLIKL SSPATLNSRV SAIALPKSCP AAGTQCLISG WGNTQSIGQN YPDVLQCLKA PILSDSVCRN AYPGQISSNM MCLGYMEGGK DSCQGDSGGP VVCNGELQGV VSWGAGCAQK GKPGVSPKVC KYVSWIQQTI AAN | |
Greater than 85% as determined by SDS-PAGE. |