missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dog Caveolin-1 Protein

A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Caveolin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9031-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16067491

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001003296.1
Caveolin-1 Protein
Research Use Only
CAV1
Recombinant Protein
Yeast
MSGGKYVDSE GHLYTVPIRE QGNIYKPNNK AMAEEMSEKQ VYDAHTKEID LVNRDPKHLN DDVVKIDFED VIAEPEGTHS FDGIWKASFT TFTVTKYWFY RLLSALFGIP MALIWGIYFA ILSFLHIWAV VPCIKSFLIE IQCISRVYSI YVHTFCDPFF EAVGKIFSNI RINMQKET
Greater than 90% as determined by SDS-PAGE.
403980
50 μg
Not determined.
Full Length
Dog
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers