missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog Epididymal secretory protein E1 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Epididymal secretory protein E1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7631-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
Epididymal secretory protein E1 Protein | |
Research Use Only | |
NPC | |
Recombinant Protein | |
E. coli | |
VHFKDCGSAV GVIKELNVNP CPAQPCKLHK GQSYSVNVTF TSNIPSQSSK AVVHGIVLGV AVPFPIPEAD GCKSGINCPI QKDKTYSYLN KLPVKNEYPS IKLVVQWMLL GDNNQHLFCW EIPVQIEG | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Full Length | |
Dog | |
His | |
Tris-based buffer, 50% glycerol |