missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dog IL-18 / IL-1F4 Protein

A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) IL-18 / IL-1F4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8263-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16043631

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001003169.1
IL-18 / IL-1F4 Protein
Research Use Only
IL18
Recombinant Protein
E. coli
YFGKLEPKLS IIRNLNDQVL FVNEGNQPVF EDMPDSDCTD NAPHTIFIIY MYKDSLTRGL AVTISVKYKT MSTLSCKNKT ISFQKMSPPD SINDEGNDII FFQRSVPGHD DKIQFESSLY KGHFLACKKE NDLFKLILKD KDENGDKSIM FTVQNKS
Greater than 90% as determined by SDS-PAGE.
403796
50 μg
Not determined.
Full Length
Dog
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers