missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 30S ribosomal protein S11 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S11, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8621-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_417756.1 | |
30S ribosomal protein S11 Protein | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
31655987 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
rpsK | |
Recombinant Protein | |
E. coli | |
AKAPIRARKR VRKQVSDGVA HIHASFNNTI VTITDRQGNA LGWATAGGSG FRGSRKSTPF AAQVAAERCA DAVKEYGIKN LEVMVKGPGP GRESTIRALN AAGFRITNIT DVTPIPHNGC RPPKKRRV |