missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 30S ribosomal protein S4 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S4, was expressed in the hosts and tags indicated.
4495.00 SEK - 27205.00 SEK
Specifications
Accession Number | NP_417755.1 |
---|---|
Gene ID (Entrez) | 32366218 |
Name | 30S ribosomal protein S4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16045571
|
enQuireBio™
QP8613-EC-10UG |
10 μg |
4550.00 SEK
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16035571
|
enQuireBio™
QP8613-EC-100UG |
100 μg |
11125.00 SEK
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16085571
|
enQuireBio™
QP8613-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065571
|
enQuireBio™
QP8613-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075571
|
enQuireBio™
QP8613-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16055571
|
enQuireBio™
QP8613-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_417755.1 | |
30S ribosomal protein S4 Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
32366218 | |
Research Use Only | |
rpsD | |
Recombinant Protein | |
E. coli | |
ARYLGPKLKL SRREGTDLFL KSGVRAIDTK CKIEQAPGQH GARKPRLSDY GVQLREKQKV RRIYGVLERQ FRNYYKEAAR LKGNTGENLL ALLEGRLDNV VYRMGFGATR AEARQLVSHK AIMVNGRVVN IASYQVSPND VVSIREKAKK QSRVKAALEL AEQREKPTWL EVDAGKMEGT FKRKPERSDL SADINEHLIV ELYSK | |
Greater than 90% as determined by SDS-PAGE. |