missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 50S ribosomal protein L30 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 50S ribosomal protein L30, was expressed in the hosts and tags indicated.
3070.00 SEK - 27205.00 SEK
Specifications
Accession Number | NP_417761.1 |
---|---|
Gene ID (Entrez) | 947797 |
Name | 50S ribosomal protein L30 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16025681
|
enQuireBio™
QP8633-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065681
|
enQuireBio™
QP8633-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16015681
|
enQuireBio™
QP8633-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16045681
|
enQuireBio™
QP8633-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16055681
|
enQuireBio™
QP8633-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16035681
|
enQuireBio™
QP8633-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_417761.1 | |
50S ribosomal protein L30 Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
947797 | |
Research Use Only | |
rpmD | |
Recombinant Protein | |
E. coli | |
MAKTIKITQT RSAIGRLPKH KATLLGLGLR RIGHTVERED TPAIRGMINA VSFMVKVEE | |
Greater than 90% as determined by SDS-PAGE. |