missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 50S ribosomal protein L33 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 50S ribosomal protein L33, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8628-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_418093.1 | |
50S ribosomal protein L33 Protein | |
Research Use Only | |
rpmG | |
Recombinant Protein | |
E. coli | |
AKGIREKIKL VSSAGTGHFY TTTKNKRTKP EKLELKKFDP VVRQHVIYKE AKI | |
Greater than 90% as determined by SDS-PAGE. |
8913847 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |