missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli ADK Protein
A DNA sequence encoding the Escherichia coli (strain K12) ADK, was expressed in the hosts and tags indicated.
3070.00 SEK - 38570.00 SEK
Specifications
Accession Number | NP_415007.1 |
---|---|
Gene ID (Entrez) | 945097 |
Name | ADK Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16016861
|
enQuireBio™
QP8857-EC-100UG |
100 μg |
11125.00 SEK
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036861
|
enQuireBio™
QP8857-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16026861
|
enQuireBio™
QP8857-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16046861
|
enQuireBio™
QP8857-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056861
|
enQuireBio™
QP8857-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066861
|
enQuireBio™
QP8857-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_415007.1 | |
ADK Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
945097 | |
Research Use Only | |
adk | |
Recombinant Protein | |
E. coli | |
MRIILLGAPG AGKGTQAQFI MEKYGIPQIS TGDMLRAAVK SGSELGKQAK DIMDAGKLVT DELVIALVKE RIAQEDCRNG FLLDGFPRTI PQADAMKEAG INVDYVLEFD VPDELIVDRI VGRRVHAPSG RVYHVKFNPP KVEGKDDVTG EELTTRKDDQ EETVRKRLVE YHQMTAPLIG YYSKEAEAGN TKYAKVDGTK PVAEVRADLE KILG | |
Greater than 90% as determined by SDS-PAGE. |