Läs mer
enQuireBio™ Recombinant E. coli Aerobic glycerol-3-phosphate dehydrogenase Protein
Produktkod. 16077190
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Förpackningsstorlek:
100µg
10µg
1mg
200µg
500µg
50µg
Produktkod. 16077190
Brand: enQuireBio™ QP7047ye50ug
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
A DNA sequence encoding the Escherichia coli (strain K12) Aerobic glycerol-3-phosphate dehydrogenase, was expressed in the hosts and tags indicated.
Specifications
Tillträdesnummer NP_417884.1
Gen-ID (Entrez) 947934
Namn Aerobic glycerol-3-phosphate dehydrogenase Protein
Kvantitet 50 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol glpD
Protein Längd Full Length
Produkttyp Recombinant Protein
Korsreaktivitet E. coli
Art Yeast
Protein Tag His
Sekvens METKDLIVIG GGINGAGIAA DAAGRGLSVL MLEAQDLACA TSSASSKLIH GGLRYLEHYE FRLVSEALAE REVLLKMAPH IAFPMRFRLP HRPHLRPAWM IRIGLFMYDH LGKRTSLPGS TGLRFGANSV LKPEIKRGFE YSDCWVDDAR LVLANAQMVV RKGGEVLTRT RATSARRENG LWIVEAEDID TGKKYSWQAR GLVNATGPWV KQFFDDGMHL PSPYGIRLIK GSHIVVPRVH TQKQAYILQN EDKRIVFVIP WMDEFSIIGT TDVEYKGDPK AVKIEESEIN YLLNVYNTHF KKQLSRDDIV WTYSGVRPLC DDESDSPQAI TRDYTLDIHD ENGKAPLLSV FGGKLTTYRK LAEHALEKLT PYYQGIGPAW TKESVLPGGA IEGDRDDYAA RLRRRYPFLT ESLARHYART YGSNSELLLG NAGTVSDLGE DFGHEFYEAE LKYLVDHEWV RRADDALWRR TKQGMWLNAD QQSRVSQWLV EYTQQRLSLA S
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 90% as determined by SDS-PAGE.