missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Aerobic glycerol-3-phosphate dehydrogenase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Aerobic glycerol-3-phosphate dehydrogenase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7047-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_417884.1 | |
Aerobic glycerol-3-phosphate dehydrogenase Protein | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
947934 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
glpD | |
Recombinant Protein | |
Yeast | |
METKDLIVIG GGINGAGIAA DAAGRGLSVL MLEAQDLACA TSSASSKLIH GGLRYLEHYE FRLVSEALAE REVLLKMAPH IAFPMRFRLP HRPHLRPAWM IRIGLFMYDH LGKRTSLPGS TGLRFGANSV LKPEIKRGFE YSDCWVDDAR LVLANAQMVV RKGGEVLTRT RATSARRENG LWIVEAEDID TGKKYSWQAR GLVNATGPWV KQFFDDGMHL PSPYGIRLIK GSHIVVPRVH TQKQAYILQN EDKRIVFVIP WMDEFSIIGT TDVEYKGDPK AVKIEESEIN YLLNVYNTHF KKQLSRDDIV WTYSGVRPLC DDESDSPQAI TRDYTLDIHD ENGKAPLLSV FGGKLTTYRK LAEHALEKLT PYYQGIGPAW TKESVLPGGA IEGDRDDYAA RLRRRYPFLT ESLARHYART YGSNSELLLG NAGTVSDLGE DFGHEFYEAE LKYLVDHEWV RRADDALWRR TKQGMWLNAD QQSRVSQWLV EYTQQRLSLA S |