missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli CS6 fimbrial subunit B Protein

A DNA sequence encoding the Escherichia coli (strain K12) CS6 fimbrial subunit B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7239-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16047830

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

WP_000913743.1
200 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
CS6 fimbrial subunit B Protein
Research Use Only
cssB
Recombinant Protein
Yeast
GNWQYKSLDV NVNIEQNFIP DIDSAVRIIP VNYDSDPKLN SQLYTVEMTI PAGVSAVKIV PTDSLTSSGQ QIGKLVNVNN PDQNMNYYIR KDSGAGKFMA GQKGSFSVKE NTSYTFSAIY TGGEYPNSGY SSGTYAGHLT VSFYSN
Greater than 92.2% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers