missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli CS6 fimbrial subunit B Protein
A DNA sequence encoding the Escherichia coli (strain K12) CS6 fimbrial subunit B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7239-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
WP_000913743.1 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
CS6 fimbrial subunit B Protein | |
Research Use Only | |
cssB | |
Recombinant Protein | |
Yeast | |
GNWQYKSLDV NVNIEQNFIP DIDSAVRIIP VNYDSDPKLN SQLYTVEMTI PAGVSAVKIV PTDSLTSSGQ QIGKLVNVNN PDQNMNYYIR KDSGAGKFMA GQKGSFSVKE NTSYTFSAIY TGGEYPNSGY SSGTYAGHLT VSFYSN | |
Greater than 92.2% as determined by SDS-PAGE. |