missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Cyclophilin A / PPIA / CYPA Protein
A DNA sequence encoding the Escherichia coli O157:H7 Cyclophilin A / PPIA / CYPA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7335-ec-50ug
Additional Details : Weight : 0.01000kg
Spezifikation
NP_312241.1 | |
Cyclophilin A / PPIA / CYPA Protein | |
Research Use Only | |
PPIA | |
Recombinant Protein | |
E. coli | |
AKGDPHVLLT TSAGNIELEL DKQKAPVSVQ NFVDYVNSGF YNNTTFHRVI PGFMIQGGGF TEQMQQKKPN PPIKNEADNG LRNTRGTIAM ARTADKDSAT SQFFINVADN AFLDHGQRDF GYAVFGKVVK GMDVADKISQ VPTHDVGPYQ NVPSKPVVIL SAKVLP | |
Greater than 90% as determined by SDS-PAGE. |
915930 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |