missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli DNA-binding protein HU-alpha Protein
A DNA sequence encoding the Escherichia coli O157:H7 DNA-binding protein HU-alpha, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7329-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_312950.1 | |
DNA-binding protein HU-alpha Protein | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
914933 | |
Greater than 94.2% as determined by SDS-PAGE. | |
Research Use Only | |
hupA | |
Recombinant Protein | |
E. coli | |
MNKTQLIDVI AEKAELSKTQ AKAALESTLA AITESLKEGD AVQLVGFGTF KVNHRAERTG RNPQTGKEIK IAAANVPAFV SGKALKDAVK |