missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Glutamate--cysteine ligase Protein

A DNA sequence encoding the Escherichia coli (strain K12) Glutamate--cysteine ligase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7387-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16058600

  • 12765.00 SEK / 200µg


Estimated Shipment
03-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_417173.1
Glutamate--cysteine ligase Protein
200 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
944881
Greater than 91.2% as determined by SDS-PAGE.
Research Use Only
gshA
Recombinant Protein
E. coli
MIPDVSQALA WLEKHPQALK GIQRGLERET LRVNADGTLA TTGHPEALGS ALTHKWITTD FAEALLEFIT PVDGDIEHML TFMRDLHRYT ARNMGDERMW PLSMPCYIAE GQDIELAQYG TSNTGRFKTL YREGLKNRYG ALMQTISGVH YNFSLPMAFW QAKCGDISGA DAKEKISAGY FRVIRNYYRF GWVIPYLFGA SPAICSSFLQ GKPTSLPFEK TECGMYYLPY ATSLRLSDLG YTNKSQSNLG ITFNDLYEYV AGLKQAIKTP SEEYAKIGIE KDGKRLQINS NVLQIENELY APIRPKRVTR SGESPSDALL RGGIEYIEVR SLDINPFSPI GVDEQQVRFL DLFMVWCALA DAPEMSSSEL ACTRVNWNRV ILEGRKPGLT LGIGCETAQF PLPQVGKDLF RDLKRVAQTL DSINGGEAYQ KVCDELVACF DNPDLTFSAR ILRSMIDTGI GGTGKAFAEA YRNLLREEPL EILREEDFVA EREASERRQQ EMEAADTEPF AVWLEKHA
Videos
SDS
Documents