missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Hygromycin-B 4-O-kinase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Hygromycin-B 4-O-kinase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7397-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
WP_000742814.1 | |
Hygromycin-B 4-O-kinase Protein | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
13905269 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
hph | |
Recombinant Protein | |
E. coli | |
MKKPELTATS VEKFLIEKFD SVSDLMQLSE GEESRAFSFD VGGRGYVLRV NSCADGFYKD RYVYRHFASA ALPIPEVLDI GEFSESLTYC ISRRAQGVTL QDLPETELPA VLQPVAEAMD AIAAADLSQT SGFGPFGPQG IGQYTTWRDF ICAIADPHVY HWQTVMDDTV SASVAQALDE LMLWAEDCPE VRHLVHADFG SNNVLTDNGR ITAVIDWSEA MFGDSQYEVA NIFFWRPWLA CMEQQTRYFE RRHPELAGSP RLRAYMLRIG LDQLYQSLVD GNFDDAAWAQ GRCDAIVRSG AGTVGRTQIA RRSAAVWTDG CVEVLADSGN RRPSTRPRAK E |