missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Lactose operon repressor Protein

A DNA sequence encoding the Escherichia coli (strain K12) Lactose operon repressor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7406-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16038710

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_414879.3
Lactose operon repressor Protein
Research Use Only
lacI
Recombinant Protein
E. coli
MKPVTLYDVA EYAGVSYQTV SRVVNQASHV SAKTREKVEA AMAELNYIPN RVAQQLAGKQ SLLIGVATSS LALHAPSQIV AAIKSRADQL GASVVVSMVE RSGVEACKAA VHNLLAQRVS GLIINYPLDD QDAIAVEAAC TNVPALFLDV SDQTPINSII FSHEDGTRLG VEHLVALGHQ QIALLAGPLS SVSARLRLAG WHKYLTRNQI QPIAEREGDW SAMSGFQQTM QMLNEGIVPT AMLVANDQMA LGAMRAITES GLRVGADISV VGYDDTEDSS CYIPPLTTIK QDFRLLGQTS VDRLLQLSQG QAVKGNQLLP VSLVKRKTTL APNTQTASPR ALADSLMQLA RQVSRLESGQ
Greater than 90% as determined by SDS-PAGE.
945007
200 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers