missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Long-chain-fatty-acid--CoA ligase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Long-chain-fatty-acid--CoA ligase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6937-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_416319.1 | |
Long-chain-fatty-acid--CoA ligase Protein | |
Research Use Only | |
fadD | |
Recombinant Protein | |
E. coli | |
MKKVWLNRYP ADVPTEINPD RYQSLVDMFE QSVARYADQP AFVNMGEVMT FRKLEERSRA FAAYLQQGLG LKKGDRVALM MPNLLQYPVA LFGILRAGMI VVNVNPLYTP RELEHQLNDS GASAIVIVSN FAHTLEKVVD KTAVQHVILT RMGDQLSTAK GTVVNFVVKY IKRLVPKYHL PDAISFRSAL HNGYRMQYVK PELVPEDLAF LQYTGGTTGV AKGAMLTHRN MLANLEQVNA TYGPLLHPGK ELVVTALPLY HIFALTINCL LFIELGGQNL LITNPRDIPG LVKELAKYPF TAITGVNTLF NALLNNKEFQ QLDFSSLHLS AGGGMPVQQV VAERWVKLTG QYLLEGYGLT ECAPLVSVNP YDIDYHSGSI GLPVPSTEAK LVDDDDNEVP PGQPGELCVK GPQVMLGYWQ RPDATDEIIK NGWLHTGDIA VMDEEGFLRI VDRKKDMILV SGFNVYPNEI EDVVMQHPGV QEVAAVGVPS GSSGEAVKIF VVKKDPSLTE ESLVTFCRRQ LTGYKVPKLV EFRDELPKSN VGKILRRELR DEARGKVDNK A | |
Greater than 90% as determined by SDS-PAGE. |
946327 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |