missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Long-chain fatty acid transport Protein
A DNA sequence encoding the Escherichia coli O157:H7 Long-chain fatty acid transport protein, was expressed in the hosts and tags indicated.
3070.00 SEK - 38570.00 SEK
Specifications
Accession Number | NP_311254.2 |
---|---|
Gene ID (Entrez) | 915676 |
Name | Long-chain fatty acid transport Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16051081
|
enQuireBio™
QP7835-EC-100UG |
100 μg |
11125.00 SEK
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071081
|
enQuireBio™
QP7835-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16061081
|
enQuireBio™
QP7835-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081081
|
enQuireBio™
QP7835-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091081
|
enQuireBio™
QP7835-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001091
|
enQuireBio™
QP7835-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_311254.2 | |
Long-chain fatty acid transport Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
915676 | |
Research Use Only | |
fadL | |
Recombinant Protein | |
E. coli | |
AGFQLNEFSS SGLGRAYSGE GAIADDAGNV SRNPALITMF DRPTFSAGAV YIDPDVNISG TSPSGRSLKA DNIAPTAWVP NMHFVAPIND QFGWGASITS NYGLATEFND TYAGGSVGGT TDLETMNLNL SGAYRLNNAW SFGLGFNAVY ARAKIERFAG DLGQLVAGQI MQSPAGKTPQ GQALAATANG IDSNTKIAHL NGNQWGFGWN AGILYELDKN NRYALTYRSE VKIDFKGNYS SDLNRVFNNY GLPIPTATGG ATQSGYLTLN LPEMWEVSGY NRVDPQWAIH YSLAYTSWSQ FQQLKATSTS GDTLFQKHEG FKDAYRIALG TTYYYDDNWT FRTGIAFDDS PVPAQNRSIS IPDQDRFWLS AGTTYAFNKD ASVDVGVSYM HGQSVKINEG PYQFESEGKA WLFGTNFNYA F | |
Greater than 90% as determined by SDS-PAGE. |