missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli LptA Protein
A DNA sequence encoding the Escherichia coli (strain K12) LptA, was expressed in the hosts and tags indicated.
3490.00 SEK - 46145.00 SEK
Specifications
Accession Number | NP_417667.1 |
---|---|
Gene ID (Entrez) | 947920 |
Name | LptA Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008260
|
enQuireBio™
QP7331-YE-100UG |
100 μg |
12410.00 SEK
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16028260
|
enQuireBio™
QP7331-YE-1MG |
1 mg |
46145.00 SEK
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018260
|
enQuireBio™
QP7331-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038260
|
enQuireBio™
QP7331-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16048260
|
enQuireBio™
QP7331-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058260
|
enQuireBio™
QP7331-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_417667.1 | |
LptA Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
947920 | |
Research Use Only | |
lptA | |
Recombinant Protein | |
Yeast | |
VTGDTDQPIH IESDQQSLDM QGNVVTFTGN VIVTQGTIKI NADKVVVTRP GGEQGKEVID GYGKPATFYQ MQDNGKPVEG HASQMHYELA KDFVVLTGNA YLQQVDSNIK GDKITYLVKE QKMQAFSDKG KRVTTVLVPS QLQDKNNKGQ TPAQKKGN | |
Greater than 90% as determined by SDS-PAGE. |