missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli LptA Protein
A DNA sequence encoding the Escherichia coli (strain K12) LptA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7331-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_417667.1 | |
LptA Protein | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
947920 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
lptA | |
Recombinant Protein | |
Yeast | |
VTGDTDQPIH IESDQQSLDM QGNVVTFTGN VIVTQGTIKI NADKVVVTRP GGEQGKEVID GYGKPATFYQ MQDNGKPVEG HASQMHYELA KDFVVLTGNA YLQQVDSNIK GDKITYLVKE QKMQAFSDKG KRVTTVLVPS QLQDKNNKGQ TPAQKKGN |