missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Mannose-6-phosphate isomerase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Mannose-6-phosphate isomerase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6374-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_416130.3 | |
Mannose-6-phosphate isomerase Protein | |
Research Use Only | |
manA | |
Recombinant Protein | |
E. coli | |
MQKLINSVQN YAWGSKTALT ELYGMENPSS QPMAELWMGA HPKSSSRVQN AAGDIVSLRD VIESDKSTLL GEAVAKRFGE LPFLFKVLCA AQPLSIQVHP NKHNSEIGFA KENAAGIPMD AAERNYKDPN HKPELVFALT PFLAMNAFRE FSEIVSLLQP VAGAHPAIAH FLQQPDAERL SELFASLLNM QGEEKSRALA ILKSALDSQQ GEPWQTIRLI SEFYPEDSGL FSPLLLNVVK LNPGEAMFLF AETPHAYLQG VALEVMANSD NVLRAGLTPK YIDIPELVAN VKFEAKPANQ LLTQPVKQGA ELDFPIPVDD FAFSLHDLSD KETTISQQSA AILFCVEGDA TLWKGSQQLQ LKPGESAFIA ANESPVTVKG HGRLARVYNK L | |
Greater than 90% as determined by SDS-PAGE. |
944840 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |