missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli NAD(P)H-flavin reductase Protein
A DNA sequence encoding the Escherichia coli (strain K12) NAD(P)H-flavin reductase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8942-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_418286.1 | |
NAD(P)H-flavin reductase Protein | |
Research Use Only | |
fre | |
Recombinant Protein | |
E. coli | |
TTLSCKVTSV EAITDTVYRV RIVPDAAFSF RAGQYLMVVM DERDKRPFSM ASTPDEKGFI ELHIGASEIN LYAKAVMDRI LKDHQIVVDI PHGEAWLRDD EERPMILIAG GTGFSYARSI LLTALARNPN RDITIYWGGR EEQHLYDLCE LEALSLKHPG LQVVPVVEQP EAGWRGRTGT VLTAVLQDHG TLAEHDIYIA GRFEMAKIAR DLFCSERNAR EDRLFGDAFA FI | |
Greater than 90% as determined by SDS-PAGE. |
948325 | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |