Learn More
enQuireBio™ Recombinant E. coli Outer-membrane lipoprotein carrier Protein
A DNA sequence encoding the Escherichia coli O9:H4 (strain HS) Outer-membrane lipoprotein carrier protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7432-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
WP_001295343.1 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
lolA | |
Recombinant Protein | |
Yeast | |
DAASDLKSRL DKVSSFHASF TQKVTDGSGA AVQEGQGDLW VKRPNLFNWH MTQPDESILV SDGKTLWFYN PFVEQATATW LKDATGNTPF MLIARNQSSD WQQYNIKQNG DDFVLTPKAS NGNLKQFTIN VGRDGTIHQF SAVEQDDQRS SYQLKSQQNG AVDAAKFTFT PPQGVTVDDQ RK |
Outer-membrane lipoprotein carrier Protein | |
10 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |