missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Parathyroid Hormone Protein
A DNA sequence encoding the Escherichia coli (strain K12) Parathyroid Hormone, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7318-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_415722.1 | |
Parathyroid Hormone Protein | |
Research Use Only | |
PTH | |
Recombinant Protein | |
E. coli | |
MTIKLIVGLA NPGAEYAATR HNAGAWFVDL LAERLRAPLR EEAKFFGYTS RVTLGGEDVR LLVPTTFMNL SGKAVAAMAS FFRINPDEIL VAHDELDLPP GVAKFKLGGG HGGHNGLKDI ISKLGNNPNF HRLRIGIGHP GDKNKVVGFV LGKPPVSEQK LIDEAIDEAA RCTEMWFTDG LTKATNRLHA FKAQ | |
Greater than 90% as determined by SDS-PAGE. |
945765 | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |