missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Peptide deformylase Protein

A DNA sequence encoding the Escherichia coli (strain K12) Peptide deformylase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6476-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16053580

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_417745.1
Peptide deformylase Protein
Research Use Only
def
Recombinant Protein
Yeast
SVLQVLHIPD ERLRKVAKPV EEVNAEIQRI VDDMFETMYA EEGIGLAATQ VDIHQRIIVI DVSENRDERL VLINPELLEK SGETGIEEGC LSIPEQRALV PRAEKVKIRA LDRDGKPFEL EADGLLAICI QHEMDHLVGK LFMDYLSPLK QQRIRQKVEK LDRLKARA
Greater than 90% as determined by SDS-PAGE.
947780
10 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions