missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli plsC Protein

A DNA sequence encoding the Escherichia coli (strain K12) plsC, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7206-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16057700

  • 8584.00 SEK / 100µg


Estimated Shipment
11-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_417490.1
plsC Protein
100 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
947496
Greater than 90% as determined by SDS-PAGE.
Research Use Only
plsC
Recombinant Protein
Yeast
MLYIFRLIIT VIYSILVCVF GSIYCLFSPR NPKHVATFGH MFGRLAPLFG LKVECRKPTD AESYGNAIYI ANHQNNYDMV TASNIVQPPT VTVGKKSLLW IPFFGQLYWL TGNLLIDRNN RTKAHGTIAE VVNHFKKRRI SIWMFPEGTR SRGRGLLPFK TGAFHAAIAA GVPIIPVCVS TTSNKINLNR LHNGLVIVEM LPPIDVSQYG KDQVRELAAH CRSIMEQKIA ELDKEVAERE AAGKV
Videos
SDS
Documents