missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli RNA-splicing ligase RtcB Protein

A DNA sequence encoding the Escherichia coli (strain K12) RNA-splicing ligase RtcB, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7212-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16067730

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_417879.1
RNA-splicing ligase RtcB Protein
Research Use Only
rtcB
Recombinant Protein
E. coli
MNYELLTTEN APVKMWTKGV PVEADARQQL INTAKMPFIF KHIAVMPDVH LGKGSTIGSV IPTKGAIIPA AVGVDIGCGM NALRTALTAE DLPENLAELR QAIETAVPHG RTTGRCKRDK GAWENPPVNV DAKWAELEAG YQWLTQKYPR FLNTNNYKHL GTLGTGNHFI EICLDESDQV WIMLHSGSRG IGNAIGTYFI DLAQKEMQET LETLPSRDLA YFMEGTEYFD DYLKAVAWAQ LFASLNRDAM MENVVTALQS ITQKTVRQPQ TLAMEEINCH HNYVQKEQHF GEEIYVTRKG AVSARAGQYG IIPGSMGAKS FIVRGLGNEE SFCSCSHGAG RVMSRTKAKK LFSVEDQIRA TAHVECRKDA EVIDEIPMAY KDIDAVMAAQ SDLVEVIYTL RQVVCVKG
Greater than 81% as determined by SDS-PAGE.
947929
10 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers