missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli RNA-splicing ligase RtcB Protein
A DNA sequence encoding the Escherichia coli (strain K12) RNA-splicing ligase RtcB, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7212-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_417879.1 | |
RNA-splicing ligase RtcB Protein | |
Research Use Only | |
rtcB | |
Recombinant Protein | |
E. coli | |
MNYELLTTEN APVKMWTKGV PVEADARQQL INTAKMPFIF KHIAVMPDVH LGKGSTIGSV IPTKGAIIPA AVGVDIGCGM NALRTALTAE DLPENLAELR QAIETAVPHG RTTGRCKRDK GAWENPPVNV DAKWAELEAG YQWLTQKYPR FLNTNNYKHL GTLGTGNHFI EICLDESDQV WIMLHSGSRG IGNAIGTYFI DLAQKEMQET LETLPSRDLA YFMEGTEYFD DYLKAVAWAQ LFASLNRDAM MENVVTALQS ITQKTVRQPQ TLAMEEINCH HNYVQKEQHF GEEIYVTRKG AVSARAGQYG IIPGSMGAKS FIVRGLGNEE SFCSCSHGAG RVMSRTKAKK LFSVEDQIRA TAHVECRKDA EVIDEIPMAY KDIDAVMAAQ SDLVEVIYTL RQVVCVKG | |
Greater than 81% as determined by SDS-PAGE. |
947929 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |